Would you like to log in again?
Fluid A Prepared Bottled Media, BD Diagnostics
Supplier: BD Diagnostics
290652 290821 290652 290821
90004-129PK 415.14 USD InStock
CA90004-128 90004-129 CA90004-138 90004-139
Fluid A Prepared Bottled Media, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
BD Prepared Bottled Media are ready-to-use formulations intended for the detection of micororganisms from pharmaceutical, food, environmental or water testing samples. They are offered in a wide variety of bottle styles and fill volumes designed to meet customer requirements.
  • Use of highest quality BBL and Difco brand dehydrated culture media for preparation
  • Conformation to current USP and EP requirements (see package labeling for details)
  • Color-coded caps and/or labeling for most products, providing easy product identification
  • Transparent labeling to allow a clear view of media and culture
Serum bottle holds up to 100 mL fill volume.
Septum/Screw Cap style has a black, phenolic screw cap with septum.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty