Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

BBL™ Fluid Thioglycolate Medium with Beef Extract, BD Life Sciences

Supplier: BD Life Sciences
BD BBL™
292333
90004-222PK 271.04 USD InStock
90004-222
BBL™ Fluid Thioglycolate Medium with Beef Extract, BD Life Sciences
Media for Microbiology Prepared Media for Microbiology
Fluid thioglycolate medium with 100 ml beef extract.

  • Shelf life: 365 days

Weight: 2.849 kg.

Packaging: Ten per pack.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty