Would you like to log in again?
Difco™ Select APS™ LB Broth Base, BD Diagnostics
Supplier: BD Diagnostics
292438 292438
90004-314EA 479.03 USD InStock
CA90004-314 90004-314
Difco™ Select APS™ LB Broth Base, BD Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Select APS™ LB Broth Base is an animal-free medium used to grow and maintain recombinant strains of Escherichia coli.
  • Excellent all-purpose growth medium
  • Manufactured from animal-free ingredients
Growth is evident by the appearance of turbidity.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty