Would you like to log in again?
Difco™ Pseudomonas Isolation Agar, BD
Supplier: BD Diagnostics
292710 292710
90004-394EA 368.15 USD InStock
90004-394 CA90004-394
Difco™ Pseudomonas Isolation Agar, BD
Media for Microbiology Dehydrated Media for Microbiology
Pseudomonas Isolation Agar is used with added glycerol in isolating Pseudomonas and differentiating Pseudomonas aeruginosa from other pseudomonads based on pigment formation.
  • Peptone provides a source of carbon and nitrogen
  • Magnesium chloride and potassium sulfate promote production of pyocyanin
  • Irgasan selectively inhibits contaminant bacteria
  • Agar is the solidifying agent
Glycerol serves as an energy source and also helps to promote pyocyanin production.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty