Would you like to log in again?
BBL™ Fluid Thioglycollate, BD
Supplier: BD Biosciences
BBL™
292769 221195 221196 299108 299803 220889 211260 299108 220889 211260 221195 221196 299502
90001-260CS 199.11 USD InStock
CA-90001-260 90001-260 90005-286 90005-176 89049-062 CA90005-176 90001-144 CA90001-144 90001-258 CA90001-258 90004-408 CA90000-132 90000-132
BBL™ Fluid Thioglycollate, BD
Media for Microbiology Dehydrated Media for Microbiology
Fluid thioglycolate is a reducing medium containing sodium thioglycolate, which reacts with molecular oxygen to keep free oxygen levels low.
  • Presence creates a redox potential in test tubes
  • Allows the differentiation of various aerobes and anaerobes
Fluid thioglycolate medium is a multipurpose, enriched, differential medium used primarily to determine the oxygen requirements of microorganisms.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty