Would you like to log in again?
Modified Letheen Agar, BD Difco™, BD Life Sciences
Supplier: BD Life Sciences
BD Difco™
263110 292847 292847 263110
90003-734EA 275.64 USD InStock
CA90003-734 90003-734 CA90004-444 90004-444
Modified Letheen Agar, BD Difco™, BD Life Sciences
Media for Microbiology Dehydrated Media for Microbiology
Modified letheen agar neutralizes quaternary ammonium compounds and is used for microbiological evaluation of cosmetics.
  • Neutralizes quaternary ammonium compounds
  • Microbiological evaluation of cosmetics
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty