Would you like to log in again?
Tryptone Azolectin Tween (TAT) Broth Base, BD Biosciences
Supplier: BD Biosciences
298410 290721 292848 298410
90004-446EA 1162.55 USD InStock
90004-446 90005-136 CA90005-136 90004-137
Tryptone Azolectin Tween (TAT) Broth Base, BD Biosciences
Media for Microbiology Additives and Supplements for Microbiology
TAT Broth Base is used for cultivating microorganisms from highly viscous or gelatinous materials.
  • Peptone provides nitrogen, vitamins, amino acids, and carbon
  • Especially adapted to the testing of cosmetics
Soy Lecithin and polysorbate 20 neutralize preservatives in the cosmetics or pharmaceutical products, allowing bacteria to grow.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty