Would you like to log in again?
Difco™ McClung Toabe Agar Base, BD Diagnostics
Supplier: BD Diagnostics
Difco™
294110 294110
90004-540EA 487.13 USD InStock
CA90004-540 90004-540
Difco™ McClung Toabe Agar Base, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
McClung Toabe Agar is used for detection and isolation of Clostridium perfringens from food samples.
  • Peptone provide nitrogenous growth nutrients
  • Dextrose is a fermentable carbohydrate
  • Phosphates form a good buffering system
  • Sodium chloride provides essential ions
  • Magnesium sulphate provides divalent cations and sulphate
With the addition of 50% egg yolk emulsion, C. perfringens and a few other Clostridium species show the lecithinase reaction. Lecithinase enzyme lyses egg yolk lecithin, producing an opaque zone of precipitation surrounding the slightly raised colonies.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty