Would you like to log in again?
BBL™ Mueller Hinton Broth, BD Life Sciences
Supplier: BD Life Sciences
BBL™
211443 296164 296164 211443
90004-658PK 433.17 USD InStock
90004-658 CA90000-194 CA-90004-658 90000-194
BBL™ Mueller Hinton Broth, BD Life Sciences
Media for Microbiology Prepared Media for Microbiology
Mueller Hinton Broth is a general-purpose medium that may be used in the cultivation of a wide variety of fastidious and nonfastidious microorganisms.
  • Casein and beef extract provide essential nitrogen and nutrients
  • Starch protects against toxic substances that may be present
  • Dextrose produced by hydrolysis is a source of energy
Mueller Hinton Broth, unadjusted, has a formula similar to that of the solid medium, but without agar, for use when fluid medium is preferred. It may be used for the general cultivation of bacteria.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty