Would you like to log in again?
BD BBL™ Prepared Tubed Diluent, Polysorbate 80 Water, BD
Supplier: BD Diagnostics
BD BBL™
296184
90004-662PK 503.63 USD InStock
90004-662
BD BBL™ Prepared Tubed Diluent, Polysorbate 80 Water, BD
Media for Microbiology Additives and Supplements for Microbiology
Polysorbate 80 water may be used to restore and/or inoculate microdilution panels.
  • On receipt, store tubes at 2 to 25 °C
  • Avoid freezing and overheating
  • Warm to room temperature before use
  • Do not open until ready to use
Polysorbate 80 is a surface active agent that is recommended for use at a 0.02% concentration in routine inoculum preparation and dispensing procedures in various microdilution systems.
Packaging: Pack containing 100, size A tubes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty