Would you like to log in again?
BBL™ Lim Broth (Todd Hewitt with CNA) For Group B Streptococcus, BD Diagnostics
Supplier: BD Life Sciences
BBL™
296266 296266
90004-686PK 286.26 USD InStock
CA90004-686 90004-686
BBL™ Lim Broth (Todd Hewitt with CNA) For Group B Streptococcus, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Lim Broth is used for the selective enrichment of group B streptococci (Streptococcus agalactiae).
  • Peptones, dextrose, and salts provide essential nutrients
  • Yeast extract is a source of B-complex vitamins
  • Dextrose stimulates hemolysin production
  • Nalidixic acid and colistin suppress growth of gram-negative bacteria
Lim Broth is used for the selective enrichment of group B streptococci (Streptococcus agalactiae), especially from genital specimens.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty