Would you like to log in again?
Difco™ BBL™ Sulfite Agar, BD Life Sciences
Supplier: BD Life Sciences
297210 297210
90004-766EA 520.35 USD InStock
CA90004-766 90004-766
Difco™ BBL™ Sulfite Agar, BD Life Sciences
Media for Microbiology Dehydrated Media for Microbiology
Sulfite Agar is used for the detection of thermophilic anaerobes producing hydrogen sulfide, particularly in foods.
  • Peptone provides carbon, nitrogen, vitamins, and minerals
  • Agar is the solidifying agent
Sulfite Agar is a recommended standard methods medium for detecting sulfide spoilage bacteria.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty