Would you like to log in again?
BBL™ Yeast Fermentation Broth, BD Diagnostics
Supplier: BD Diagnostics
BBL™
297301
90004-824PK 75.36 USD InStock
90004-824
BBL™ Yeast Fermentation Broth, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Yeast fermentation media for the isolation of tubercle fungi.
  • With sucrose or maltose and Durham's tube
  • Sterile
  • Ready-to-use
The product includes a surface area of 5.4 cm for cultivation.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty