Would you like to log in again?
BBL™ Prepared Tubed Diluent, Water, BD Diagnostics
Supplier: BD Diagnostics
BBL™
297345
90004-848PK 241.21 USD InStock
90004-848
BBL™ Prepared Tubed Diluent, Water, BD Diagnostics
Media for Microbiology Additives and Supplements for Microbiology
The water in these tubes is purified (deionised) water that is ready and convenient for use as a diluent or suspending medium.
Water has been recommended for numerous uses in the microbiology laboratory. It is utilized in serial dilutions for test cultures, in preparing stock solutions of reagents and in the preparation of the base medium for yeast assimilation studies.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty