Would you like to log in again?
BBL™ Trypticase™ Soy Broth with 20% Glycerol, BD Diagnostics
Supplier: BD Diagnostics
297352 297808 296346
90005-020PK 256.27 USD InStock
90005-020 90004-850 90004-700
BBL™ Trypticase™ Soy Broth with 20% Glycerol, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Trypticase soy broth, a nutritious medium supplemented with glycerol is used as a maintenance medium for the preservation of bacterial culture.
Enzymatic digests of protein substrates act as protective colloids
Bacterial stock cultures frozen and stored at –20 °C or below will remain viable for several months, and some may remain viable for years.
Glycerol is a cryoprotective agent that provides intracellular and extracellular protection against freezing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty