Would you like to log in again?
BBL™ Selective 7H11 (Mitchison) Agar Slants, BD
Supplier: BD Life Sciences
297639 297639
90004-926PK 1269.68 USD InStock
90004-926 CA-90004-926
BBL™ Selective 7H11 (Mitchison) Agar Slants, BD
Media for Microbiology Prepared Media for Microbiology
Selective 7H11 Agar Slants are used in qualitative procedures for isolation and cultivation of mycobacteria, especially Mycobacterium tuberculosis, from clinical and nonclinical specimens.
  • Antimicrobial agents added to reduce the need for decontamination procedures
  • Supplemented pancreatic digest of casein
  • Pyruvate enhances the recovery of M. bovis
The Selective 7H11 Agar is 7H11 Agar modified by the addition of four antimicrobial agents: polymyxin B, carbenicillin, amphotericin B, and trimethoprim lactate.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty