Would you like to log in again?
BBL™ Fluid Thioglycollate Medium, Enriched, BD Diagnostics
Supplier: BD Life Sciences
BD BBL™
297642 297642
90004-930PK 305.7 USD InStock
CA-90004-930 90004-930
BBL™ Fluid Thioglycollate Medium, Enriched, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Fluid thioglycollate medium enriched are general-purpose media used in qualitative procedures for the cultivation of fastidious, as well as nonfastidious microorganisms.
  • Used in the isolation and cultivation slow growing obligately anaerobic microorganismls
Media supplemented with sodium bicarbonate or a marble chip is used to prepare a standardized inoculum by the growth method for antimicrobial susceptibility testing of anaerobic bacteria.
Fluid thioglycollate medium, enriched is BBL™ fluid thioglycollate medium supplemented with vitamin K and hemin.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty