Would you like to log in again?
BBL™ Sabouraud Dextrose Agar with Chloramphenicol and Cycloheximide, BD
Supplier: BD Biosciences
BBL™
297649
90004-936PK 62.76 USD InStock
90004-936
BBL™ Sabouraud Dextrose Agar with Chloramphenicol and Cycloheximide, BD
Media for Microbiology Dehydrated Media for Microbiology
This medium is used for the cultivation of dermatophytes and other pathogenic and nonpathogenic fungi from clinical and non-clinical specimens.
  • Low pH range ~5.6
  • Peptone medium supplemented with dextrose
Developed by medium developed by Sabouraud for the cultivation of dematophytes. Favorable for the growth of fungi, especially dermatophytes, and inhibitory to contaminating bacteria in clinical specimens.
Addition of antimicrobial agents improves the recovery of pathogenic fungi from specimens heavily contaminated with bacteria and saprophytic fungi.
Chloramphenicol is a broad-spectrum antibiotic inhibitory to a wide range of gram-negative and gram-positive bacteria. Cycloheximide is an antifungal agent that is primarily active against saprophytic fungi and does not inhibit yeasts or dermatophytes.
Store at 2 to 8 °C.
Caution: Do not use medium if it shows evidence of microbial contamination, discoloration, drying, or other signs of deterioration.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty