Would you like to log in again?
Lowenstein-Jensen Medium Slants, Prepared Media in Tubes, BD Diagnostics
Supplier: BD Diagnostics
297653 221388 297703
90004-940PK 621.23 USD InStock
CA-90004-966 CA-90001-346 90004-940
Lowenstein-Jensen Medium Slants, Prepared Media in Tubes, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Media for the isolation and identification of microorganisms.
  • Ideal for cultures requiring prolonged incubation
  • Made of borosilicate glass
  • Securely fitted screw caps to prevent loss of moisture
Media for the isolation and identification of microorganisms. Tubed media are ideal for cultures requiring prolonged incubation, since the caps can be closed tightly to prevent dehydration and subsequent inhibition of growth. Long-necked, rubber-lined screw caps eliminate danger of contamination. Larger-sized tubes with wider diameters permit easy inoculation and handling of media. Tubes are made of borosilicate glass and have securely fitted screw caps to prevent loss of moisture. Caps contain a resilient rubber liner that resists displacement even during autoclaving.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty