Would you like to log in again?
BBL™ Sabouraud Brain Heart Infusion Agar, Slants, BD
Supplier: BD Life Sciences
BBL™
297252 297685 297691
90004-956PK 505.61 USD InStock
90004-956 90004-954 90004-796
BBL™ Sabouraud Brain Heart Infusion Agar, Slants, BD
Media for Microbiology Prepared Media for Microbiology
Sabouraud Brain Heart Infusion Agar Slants are used in qualitative procedures for the selective isolation and cultivation of pathogenic fungi from clinical and nonclinical specimens.
  • Peptones and infusions provide amino acids and nitrogen
  • Dextrose is an energy source
  • Sodium chloride provides essential electrolytes
  • Disodium phosphate buffers the medium to maintain the pH
  • Agar is the solidifying agent
Chloramphenicol is a broad-spectrum antibiotic that inhibits a wide range of gram-negative and gram-positive bacteria. Gentamicin is an aminoglycoside antibiotic that inhibits the growth of gram-negative bacteria.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty