Would you like to log in again?
BBL™ Saline, Normal, BD
Supplier: BD Life Sciences
BBL™
221819 221819 297753 297815 297815
90005-030PK 187.48 USD InStock
CA-90001-538 90005-030 90001-538 90004-992 CA-90005-030
BBL™ Saline, Normal, BD
Media for Microbiology Prepared Media for Microbiology
Normal saline is used in procedures that require the use of an isotonic diluent.
  • Ready to use
  • Provide ensured cell integrity
  • Used with a vast array of procedures
An isotonic diluent may be used for dilution of bacterial cells to provide a concentration suitable for microscopic observation, determination of cell numbers, analysis for genetic or metabolic properties, washing cells preparatory to study, or preparation of standardized inocula.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty