Would you like to log in again?
BBL™ Brain Heart Infusion, BD Biosciences
Supplier: BD Biosciences
BBL™
211059 211060 221812 299070 221778 220837 221812 221785 211059 221813 299070
90001-136CS 198.69 USD InStock
CA90001-530 90001-136 90001-533 90005-160 90000-060 90001-514 CA90005-160 CA90000-060 90000-062 90001-519 90001-530
BBL™ Brain Heart Infusion, BD Biosciences
Media for Microbiology Dehydrated Media for Microbiology
Brain Heart Infusion (BHI) broth is a general-purpose liquid medium for the growth of a wide variety of bacterial and fungal species.
  • Brain and heart infusions and peptones provide nutrients
  • Supports the growth of fastidious and nonfastidious microorganisms
BHI Broth is used for the cultivation of a wide variety of microorganisms, including bacteria, yeasts, and molds.
Brain heart infusion, modified differs from other formulations by the quantities of the ingredients and the substitution of pancreatic digest of casein for pancreatic digest of gelatin.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty