Would you like to log in again?
Potato Dextrose Agar, BD Diagnostics
Supplier: BD
299906
90005-300PK 384.31 USD InStock
90005-300
Potato Dextrose Agar, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Potato dextrose agar is used for the cultivation and enumeration of yeasts and molds.
Used for sterility testing, non sterile product testing, food, dairy and water testing.
Certifications: Conforms to EP/USP.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty