Would you like to log in again?
BBL™ mEI Agar, BD
Supplier: BD Life Sciences
215045 215047 215045
90006-320CS 2102.08 USD InStock
90006-320 CA90006-316 90006-316
BBL™ mEI Agar, BD
Media for Microbiology Prepared Media for Microbiology
mEI Agar is a selective medium used for the chromogenic detection and enumeration of enterococci in water by the single-step membrane filtration technique.
  • Peptone supplies nitrogen and carbon compounds
  • Sodium chloride maintains osmotic equilibrium
  • Esculin is hydrolyzed to form esculetin and dextrose
  • Sodium azide inhibits gram-negative bacteria
  • Yeast extract provides vitamins, amino acids, and trace elements
  • Agar is the solidifying agent
A wide range of sample volumes or dilutions can be tested by this single-step MF procedure for the detection and enumeration of enterococci in potable, fresh, estuarine, marine, and shellfish-growing waters.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty