Would you like to log in again?
BD BBL™ CHROMagar™ Orientations, BD Diagnostics
Supplier: BD
254102 215081
90006-346PK 358.2 USD InStock
90006-346 90008-924
BD BBL™ CHROMagar™ Orientations, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
BBL™ CHROMagar™ Orientation is a nonselective medium for the isolation, differentiation, and enumeration of urinary tract pathogens.
  • Peptones provide a source of essential nutrients
  • Chromogens release colored compounds upon degradation
  • Proteus swarming is partially to completely inhibited
The chromogen mix consists of artificial substrates (chromogens), which release differently colored compounds upon degradation by specific microbial enzymes, thus assuring the differentiation of certain species or the detection of certain groups of organisms, with only a minimum of confirmatory tests.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty