Would you like to log in again?
BD BBL™ Group A Selective Strep Agar with 5% Sheep Blood Prepared Media Stacker™ Plates, BD Diagnostics
Supplier: BD Diagnostics
221780 221779 221780 221779
90006-464PK 236.33 USD InStock
CA90006-462 90006-464 90006-463 CA-90006-464
BD BBL™ Group A Selective Strep Agar with 5% Sheep Blood Prepared Media Stacker™ Plates, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Prepared plated media are designed for the isolation of microorganisms from samples or specimens and come in 100 x 10 mm or 100 x 15 mm petri-style configurations.
  • Plates contain a single medium
  • Interlocking, stackable design to minimize sliding or sample disruption
  • Ready for immediate use
Store at 2 to 8 °C (36 to 46 °F).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty