Would you like to log in again?
BD BBL™ Test Tube Agglutination Viewer, BD Diagnostics
Supplier: BD Diagnostics
BD BBL™
420630 420630
90006-592EA 990.55 USD InStock
CA63020-066 90006-592
BD BBL™ Test Tube Agglutination Viewer, BD Diagnostics
Magnifiers
Ideal for use in blood typing, cross matching, flocculation, serodiagnostic tests, and other agglutinations
Viewer provides an easy and accurate reading of the undersurface of test tube contents. Sample contents are illuminated from above by a 5W bulb and from below by a magnifying mirror. Replacement bulbs available separately.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty