Would you like to log in again?
Enterobacterial Enrichment (EE) Broth Mossel, BD Life Sciences
Supplier: BD Life Sciences
256620 256620 292627 292627
90003-524EA 431.7 USD InStock
CA90003-524 90003-524 90007-886 CA90007-886
Enterobacterial Enrichment (EE) Broth Mossel, BD Life Sciences
Media for Microbiology Prepared Media for Microbiology
EE Broth Mossel is used for selective enrichment of Enterobacteriaceae in bacteriological examination of foods.
  • Peptones provide nitrogen, vitamins, and amino acids
  • Dextrose is a carbon source
  • Disodium phosphate and monopotassium phosphate are buffering agents
  • Brilliant green and oxgall are selective agents
EE Broth Mossel is listed in the USP as one of the recommended media for the isolatoin of bile-tolerant gram-negative bacteria from nonsterile pharmaceutical products.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty