Would you like to log in again?
BD BBL™ Brain Heart CC Agar with Sheep Blood, Prepared Media Stacker™ Plates, BD Diagnostics
Supplier: BD Diagnostics
BD BBL™
296178
90008-410PK 134.84 USD InStock
90008-410
BD BBL™ Brain Heart CC Agar with Sheep Blood, Prepared Media Stacker™ Plates, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Brain heart infusion agar with chloramphenicol, cycloheximide and sheep blood is a selective medium used for the isolation of pathogenic fungi from specimens heavily contaminated with bacteria and saprophytic fungi.
  • Ready for immediate use
  • Serves as the base for enriched and more selective media supplemented with sheep blood and antibiotics.
With 10% sheep blood, it is used to isolate systemic fungi that may grow poorly on the nonenriched medium. The presence of the antimicrobial agents, cycloheximide and/or chloramphenicol and, in modified formulations, gentamicin, penicillin and streptomycin, inhibits the growth of a wide variety of bacteria and fungi and enhances the isolation of pathogenic fungal species.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty