Would you like to log in again?
BBL™ HBT Bilayer Medium, BD
Supplier: BD Life Sciences
BBL™
297884
90008-708PK 111.13 USD InStock
90008-708
BBL™ HBT Bilayer Medium, BD
Media for Microbiology Prepared Media for Microbiology
HBT (Human Blood Tween) Bilayer Medium is a selective and differential medium used in the primary isolation and presumptive identification of G. vaginalis from clinical specimens.
  • Package of 10 plates
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty