Would you like to log in again?
BBL™ Rapid Urea Broth, BD Diagnostics
Supplier: BD Life Sciences
298330 298330
90008-756PK 207.66 USD InStock
CA90008-756 90008-756
BBL™ Rapid Urea Broth, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Rapid Urea Broth is used for the presumptive identification of Helicobacter pylori in gastric antral biopsy specimens.
  • Activity of urease is primarily due to H. pylori
  • Urease enzyme renders urease broth alkaline
  • Presense of urease changes phenol red indicator
The preformed urease enzyme present in biopsy specimens renders the urease broth alkaline and changes the phenol red indicator in the medium to a pink-red color.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty