Would you like to log in again?
BD™ Fluid Thioglycollate Medium Bottled Media with 0.5% P80, BD Diagnostics
Supplier: BD Diagnostics
BD™
257365
90009-033PK 432.17 USD InStock
90009-033
BD™ Fluid Thioglycollate Medium Bottled Media with 0.5% P80, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Fluid thioglycollate medium with 0.5% polysorbate 80 is used for testing oils or materials containing lecithin in sterile fill rooms.
Packaging: Packaged in two Stericlin® bags before autoclaving, providing a sterile outer surface of the containers.
Caution: For laboratory use only.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty