Would you like to log in again?
Tryptic Soy Broth with 0.5% Polysorbate 80, BD Diagnostics
Supplier: BD Diagnostics
257366
90009-035PK 525.7 USD InStock
90009-035
Tryptic Soy Broth with 0.5% Polysorbate 80, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Tryptic soy broth with polysorbate 80 media are used for sterility testing of specimens containing oils or lecithin.
  • General purpose medium
  • Single wrapped, septum
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty