Would you like to log in again?
Complete IV One-Step Phosphoric/Citric Acid Cleaner/Descaler, CANI
Supplier: CANI
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CANI Complete
CAN-COM4-30 CAN-COM4-15 CAN-COM4-1 CAN-COM4-5 CAN-COM4-2.5 CAN-COM4-5 CAN-COM4-55 CAN-COM4-2.5 CAN-COM4-55 CAN-COM4-15 CAN-COM4-275 CAN-COM4-30 CAN-COM4-1 CAN-COM4-275
93001-150EA 474.9 USD InStock
93001-150 93001-140 CA93001-144 93001-146 CA93001-146 93001-148 CA93001-150 93001-142 CA93001-140 CA93001-142 93001-144 93001-138 CA93001-148 CA93001-138
Complete IV One-Step Phosphoric/Citric Acid Cleaner/Descaler, CANI
Cleaning Agents Detergents and Surface Cleansers
Formulated for the research, pharmaceutical, and biotech industries
Cleaner is ideal for animal cage wash applications and will clean and descale in just one wash cycle, eliminating the use of an alkaline detergent and a full wash cycle, improving throughput, and reducing water consumption. Cleaner is also effective in pharmaceutical/biotech process cleaning and derouging. Features a low-foam formulation that will not cavitate recirculation pumps. Cleaner’s free-rinsing design provides a clean, residue-free surface, and the excellent hard water tolerance eliminates mineral deposits. The surfactant system remains effective across a wide temperature and pH range, even during pH neutralization prior to discharge, thus reducing redeposition of process residues.
Effectively removes a wide range of process and research residues, such as inorganic salts and scales, particulate carbon and urine scales, fecal matter, oils, and proteins
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty