Would you like to log in again?
CP-700 Alkaline Detergent, CANI
Supplier: CANI
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CANI CP
CAN-700-1 CAN-700-1 CAN-700-15 CAN-700-15 CAN-700-30 CAN-700-275 CAN-700-275 CAN-700-30 CAN-700-55 CAN-700-55 CAN-700-2.5 CAN-700-5 CAN-700-2.5 CAN-700-5
93001-160EA 1946.54 USD InStock
93001-160 CA93001-160 93001-162 CA93001-154 93001-156 93001-158 CA93001-156 93001-152 CA93001-162 93001-164 CA93001-152 93001-154 CA93001-164 CA93001-158
CP-700 Alkaline Detergent, CANI
Cleaning Agents Detergents and Surface Cleansers
Low-foaming, concentrated liquid alkaline detergent used to clean process and research equipment, and laboratory glassware and plastics
Ideal for pharmaceutical, biotechnology, cosmetic, medical device, and food and beverage facilities. Detergent removes a wide range of process and product residues, including proteins, organics, many excipients, and fine chemicals. Detergent is free-rinsing and phosphate-free.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty