Would you like to log in again?
CP-710 Non-Hazardous Alkaline Detergent, CANI
Supplier: CANI
CANI CP
CAN-710-55 CAN-710-15 CAN-710-5 CAN-710-30 CAN-710-15 CAN-710-2.5 CAN-710-275 CAN-710-30 CAN-710-5 CAN-710-2.5 CAN-710-1 CAN-710-275 CAN-710-1 CAN-710-55
93001-310EA 13489.9 USD InStock
93001-310 93001-312 CA93001-310 CA93001-316 CA93001-306 93001-308 CA93001-308 93001-314 CA93001-312 93001-304 93001-316 CA93001-314 93001-306 CA93001-304
CP-710 Non-Hazardous Alkaline Detergent, CANI
Cleaning Agents Detergents and Surface Cleansers
Concentrated liquid formulated for the pharmaceutical, biotechnology, cosmetic, and medical device industries
Effectively removes a wide range of process and research residues, including serum, proteins, organics, and oils. Safe for use on aluminum, galvanized and soft metals, glass, stainless steel, and a wide variety of plastics. Ideal for laboratory animal cage wash applications at 0.2–1% use dilution. Designed to be low-foaming at all temperatures for easy rinsing and will not cavitate recirculation pumps. Free-rinsing formulation provides clean, residue-free surfaces. Chelants provide excellent hard water tolerance to eliminate mineral deposits.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty