Would you like to log in again?
CP-811 Emulsifying Detergent/Additive, CANI
Supplier: CANI
CANI CP
CAN-811-2.5 CAN-811-5 CAN-811-5 CAN-811-15 CAN-811-275 CAN-811-30 CAN-811-1 CAN-811-2.5 CAN-811-55 CAN-811-1 CAN-811-55 CAN-811-15 CAN-811-30 CAN-811-275
93001-390CS 456.19 USD InStock
93001-390 CA93001-390 93001-392 CA93001-396 93001-398 93001-388 CA93001-386 CA93001-398 CA93001-388 CA93001-392 93001-394 93001-396 CA93001-394 93001-386
CP-811 Emulsifying Detergent/Additive, CANI
Cleaning Agents Detergents and Surface Cleansers
Phosphate-free, neutral pH liquid detergent and additive effectively removes a wide range of residues and is particularly effective against oils, creams, grease, petrolatums, and waxes
When used alone or with compatible alkaline cleaners, the detergent’s powerful emulsifiers and enhanced low-foam surfactant blend improve cleaning and rinsing, making it ideal for recirculating spray washers, pharmaceutical/biotech process cleaning, cosmetic process cleaning, and laboratory glassware washing. Detergent rinses completely for a clean, residue-free surface.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty