Would you like to log in again?
Millennium Citrus Cleaners, CANI
Supplier: CANI
Millennium
CAN-MCTR-1 CAN-MCTR-15 CAN-MCTR-5 CAN-MCTR-275 CAN-MCTR-30 CAN-MCTR-275 CAN-MCTR-55 CAN-MCTR-2.5 CAN-MCTR-1 CAN-MCTR-15 CAN-MCTR-30 CAN-MCTR-55 CAN-MCTR-2.5 CAN-MCTR-5
93001-442EA 1257.78 USD InStock
CA93001-440 93001-442 CA93001-442 93001-444 93001-450 93001-440 CA93001-450 CA93001-448 CA93001-438 CA93001-444 93001-446 CA93001-446 93001-448 93001-438
Millennium Citrus Cleaners, CANI
Cleaning Agents Detergents and Surface Cleansers
Highly concentrated cleaners designed to cut through the toughest soil conditions, and leave surfaces sparkling clean with a fresh, pleasant citrus odor
Cleaner is available in general-purpose and low-foaming formulations. The general-purpose cleaners contain an effective buffering agent combined with natural citrus oils to maintain cleaning strength under heavy soil loads. Cleaners also contain a fast-acting, powerful solubilizing agent that dissolves and suspends any soil, allowing it to be wiped or rinsed away completely. The low-foaming cleaners can be used as a detergent additive in immersion, pressure washing, and circulation cleaning for the cosmetic, pharmaceutical, medical device, and food and beverage industries.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty