Would you like to log in again?
Sani 512 No-Rinse Quat Sanitizer/Disinfectant, CANI
Supplier: CANI
CAN-S512-30 CAN-S512-55 CAN-S512-15
93001-490EA 838 USD InStock
93001-490 93001-498 93001-496
Sani 512 No-Rinse Quat Sanitizer/Disinfectant, CANI
Cleaning Agents Disinfectants
An effective sanitizer, disinfectant, and deodorant for use on countertops, eating utensils, drinking glasses, or any hard inanimate, nonporous surface
Will sanitize at a use dilution of 7.4mL (0.25oz.) per 3.8L (1gal.) of water. No rinsing is required when the disinfectant is used on previously cleaned hard surfaces. The compounds must always be used at dilutions and according to applicable directions provided on the EPA registered label. Effective against salmonella choleraesuis, staphylococcus aureus, and pseudomonas aeruginos.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty