Would you like to log in again?
Septa, Fluoropolymer Resin Silicone, Qorpak®
Supplier: Qorpak
CAP-00513 CAP-00515 CAP-00563
11311-628CS 75.61 USD InStock
11311-628 93001-762 16151-690
Septa, Fluoropolymer Resin Silicone, Qorpak®
Septa
Resist coring and provide great resealability.
Ideal for when multiple injections are required. Septa maintain PTFE resistance until punctured, then have silicone compatibility.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty