Would you like to log in again?
Vacuum Filtration Luer, DWK Life Sciences
Supplier: DWK Life Sciences
205100-2440 205100-1420
93001-856EA 150.9 USD InStock
93001-856 93001-860
Vacuum Filtration Luer, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
Use this adapter with disposable syringe cartridges for easy sample concentration and to eliminate the cleaning of glass frits and funnels. Designed to reduce cross-contamination.
  • Features top luer fitting and integral vacuum connection
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Ordering information: Includes drip tip. Plastic valves with luer fittings are available separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty