Would you like to log in again?
ERGOMAT® Nitro Ergonomic Floor Mat, ERGOMAT
Supplier: Ergomat
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Ergomat®
ENT0406 ENT0218 ENT0208 ENT0408 ENT0203 ENT0413 ENT0209 ENT0210 ENT0306 ENT0313 ENT0204 ENT0212 ENT0308 ENT0414 ENT0219 ENT0320 ENT0418 ENT0216 ENT0404 ENT0319 ENT0317 ENT0409 ENT0307 ENT0411 ENT0410 ENT0213 ENT0420 ENT0315 ENT0305 ENT0215 ENT0316 ENT0214 ENT0211 ENT0407 ENT0415 ENT0310 ENT0207 ENT0405 ENT0309 ENT0312 ENT0416 ENT0304 ENT0417 ENT0217 ENT0314 ENT0412 ENT0205 ENT0419 ENT0303 ENT0220 ENT0318 ENT0206 ENT0311
94000-186EA 1053.55 USD InStock
94000-186 94000-142 94000-140 94000-184 94000-182 94000-180 94000-148 94000-146 94000-188 94000-144 94000-152 94000-196 94000-194 94000-150 94000-192 94000-190 94000-116 94000-158 94000-114 94000-112 94000-156 94000-110 94000-154 94000-198 94000-118 94000-120 94000-164 94000-162 94000-160 94000-128 94000-204 94000-126 94000-202 94000-124 94000-168 94000-200 94000-122 94000-166 94000-208 94000-206 94000-174 94000-130 94000-172 94000-170 94000-138 94000-214 94000-136 94000-212 94000-178 94000-134 94000-210 94000-132 94000-176
ERGOMAT® Nitro Ergonomic Floor Mat, ERGOMAT
Floor Mats Traction and Anti-Fatigue Floor Mats
ERGOMAT Nitro mats are specifically designed to provide superior traction in excessively wet or oily working environments without sacrificing ergonomic comfort
Nitril rubber offers resistance to routine exposures between pH 2-9.5. The Nitro is nonporous and will not expand or swell like other materials when exposed to oils. Mats are latex free and suitable for use in ISO Class 5 (Class 100) cleanrooms.
These durable virgin nitril rubber mats have a unique peaked cone profile which grabs footwear in demanding and slippery environments.
Ordering information: ERGOMAT Nitro mats can be provided in any shape and size, and come with a 3 year manufacturer’s warranty. Contact your VWR representative for additional information.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty