Would you like to log in again?
HPLC Remote Wall Bracket Kits, Justrite®
Supplier: Justrite
28150 28151
94000-346EA 1116.08 USD InStock
94000-346 94000-344
HPLC Remote Wall Bracket Kits, Justrite®
Wall Hooks
Ideal for applications utilizing safety disposal cans with disconnects.
HPLC filters can often be attached directly to the vapor release fitting on disposal cans. However, when this option is unavailable, remote wall brackets allow filters to be placed in a convenient and remote location. Disconnect fitting is valved to provide a safe, fully closed system that prevents vapor release when changing filters. Fittings accommodate 9.5 mm (3/8") O.D. tubing.
Delivery information: Bracket kits consist of a female quick disconnect fitting, a type 304 stainless steel holder, and two self-tapping mounting screws. Tubing not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty