Would you like to log in again?
HPLC Poly Manifolds for Quick-Disconnect Safety Disposal Cans, Justrite®
Supplier: Justrite
28177 28189 28179 28178 28188
94000-350EA 430.43 USD InStock
94000-350 94000-348 94000-356 94000-354 94000-352
HPLC Poly Manifolds for Quick-Disconnect Safety Disposal Cans, Justrite®
Manifolds
Manifolds enable up to six waste lines (per manifold) from HPLC processes to be fed into a single intake fitting.
  • Equipped with six 1/4-28 ports, one 9.5 mm (3/8") NPT port
  • Option of one 316 stainless steel quick disconnect, one polypropylene quick disconnect, or 316 stainless steel stacker connector
  • Six ports tapped to accommodate 1/4-28 threads of compression fittings
The compression fittings accommodate 1.6 mm (1/16") or 3.2 mm (1/8") O.D tubing sizes. Tefzel® fittings are approximately 2.5 cm (1") long and include ferrules for a compression fit to the waste line.
The manifold with stainless steel stackable connection enables attachment on top of either the stainless steel or polypropylene quick disconnect manifolds. This addition allows up to a total of 12 waste lines to be fed to a disposal can.
Certifications: Safety cans with manifold installed are OSHA compliant and meet NFPA Code 30.
Ordering information: Manifolds include caps for sealing off the unused ports and reducing vapor release. Tubing not included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty