Would you like to log in again?
CSM Glovebox Gloves, Honeywell Safety
Supplier: Honeywell Safety
10Y3032A/9Q Y154A/10 Y154A/7 Y154A/8 10Y1532A/9Q 10Y1532A/9Q Y154A/9
89186-130PR 327.52 USD InStock
CA10746-664 89186-130 94000-888 94000-886 89186-134 89186-132 89186-136
CSM Glovebox Gloves, Honeywell Safety
Gloves
CSM glove has outstanding mechanical properties to protect against abrasion, tears, and ozone and oxidizing chemicals.
  • Multi-dipped construction
  • Easily cleaned
  • Unsupported
The white surface is specially designed to easily identify contamination. CSM material provides superior protection, while also having an inherently soft and comfortable feel for greater worker comfort.
Certifications: ANSI/ISEA 105 - ASTM D3389-05 Abrasion Test. ANSI/ISEA 105 - ASTM D412 - Physical properties. ANSI/ISEA 105 - ASTM F1790-97 Cut Test. EN 374 - Permiation and breakthrough time. EN 388 - Mechanical risks from abrasion, cut, tear and puncture. EN 420 glove standards. OSHA Standard -29 CFR 1915.157.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty