Would you like to log in again?
UltraSoft Tile-Top Anti-Fatigue Mat with Anti-Microbial Sponge, Wearwell®
Supplier: WEARWELL
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
419.78X4X60AMBL 419.78X2X3AMCH 419.78X2X60AMBL 419.78X2X60AMBK 419.78X3X60AMGY 419.78X3X5AMBK 419.78X4X60AMBK 419.78X3X60AMCH 419.78X3X60AMBL 419.78X3X5AMBL 419.78X2X3AMGY 419.78X2X3AMBL 419.78X2X60AMCH 419.78X3X5AMGY 419.78X4X60AMCH 419.78X3X60AMBK 419.78X2X60AMGY 419.78X4X60AMGY 419.78X3X5AMCH 419.78X2X3AMBK
94001-040EA 609.18 USD InStock
94001-040 94001-050 94001-020 94001-042 94001-052 94001-030 94001-026 94001-048 94001-036 94001-028 94001-038 94001-044 94001-022 94001-054 94001-032 94001-024 94001-046 94001-034 94001-056 94001-018
UltraSoft Tile-Top Anti-Fatigue Mat with Anti-Microbial Sponge, Wearwell®
Floor Mats Traction and Anti-Fatigue Floor Mats
Combats fatigue while providing a top to bottom anti-microbial solution.
  • Premium Anti-Microbial sponge
  • Non-porous, Anti-Microbial surface
  • 2X the resilience and 3X the life of look-alikes
  • Maximized standing area
  • Lay-flat design
UltraSoft Tile-Top AM doubles down on combating fatigue while providing a top to bottom anti-microbial solution. An anti-microbial top surface provides top level defense and is backed with an anti-microbial Nitricell® sponge. Twice as thick, it absorbs repeated compression and bounces back to its original form better than any PVC sponge alternative. Tile-Top AM is a game changer compared to typical tile-top mats for teams in labs, pharmacies, biotechnology, and pharmaceutical manufacturing. Ideally suited for use in dry environments.
Certifications: Meets ASTM F1677. Dry: COF = 0.78. Meets ASTM D395-98/20%.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty