Would you like to log in again?
ProKlenz® Booster Detergent Additives
Supplier: Steris
ProKlenz®
160905 1609D9 1609D3 160901 160901 160905
89401-558EA 8832.72 USD InStock
89401-558 CA11007-754 CA11007-752 94001-108 89401-560 94001-110
ProKlenz® Booster Detergent Additives
Cleaning Agents Detergents and Surface Cleansers
ProKlenz® Booster Detergent Additive is a proprietary blend of surfactants, an oxidizing agent and other performance-enhancing ingredients that provide multiple cleaning mechanisms. This product removes a wide range of process residues and is ideal for use in CIP, COP, and manual applications.
  • Productivity - Enhances cleaning performance of other detergents or works individually with its superior blend of surfactants and oxidizing chemistry
  • An excellent product for tablet coating removal in pharmaceutical and dietary supplement manufacturing facilities
  • Easy to validate - Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports
  • Cost savings - Reduces water consumption and cleaning time due to superior rinsing and low foaming characteristics (above 50 °C) when used in clean-in-place (CIP), clean-out-of-place (COP), and manual applications
  • Globally availability - phosphate free, and manufactured with biodegradable ingredients to support harmonization of cleaning processes
  • Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility
ProKlenz® Booster high performance detergent additive can resolve your most challenging process cleaning challenges by greatly enhancing the performance of other detergents with a state-of-the-art surfactant system and oxidizing chemistry.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty