Would you like to log in again?
ChemSplash® 1 Chemical Splash Coveralls, Elastic Wrist, Open Ankle, Taped Seams, Elastic Back, International Enviroguard™
Supplier: International Enviroguard
7012YT-2XL 7012YT-XL 7012YT-L 7012YT-M 7012YT-4XL 7012YT-3XL
94001-172CS 185.94 USD InStock
94001-172 94001-174 94001-170 94001-168 94001-164 94001-166
ChemSplash® 1 Chemical Splash Coveralls, Elastic Wrist, Open Ankle, Taped Seams, Elastic Back, International Enviroguard™
Disposable Garments
Disposable Coverall designed to protect against chemical splash and toxic particles.
  • Proprietary hybrid membrane fabric helps to keep wearer cool and avoid heat stress
  • Taped seams improve protection vs coveralls made with serged seams
  • Contrasting red stripe to quickly distinguish this taped-seam garment from serged seam coveralls
Roomy, seamless shoulder area resists tears and provides more protection in key exposure area. Wider, gusseted crotch gives wearer more room to move in all directions without ripping fabric. Elastic back stretches with wearer for more comfort and better tear resistance. Storm flap over zipper with pressure sensitive tape adds an extra layer of protection. High moisture vapor transfer rate improves wearer comfort.
Recommended applications include manufacturing, emergency response, environmental clean-up, and more.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty