Would you like to log in again?
KleenGuard™ A20 Breathable Particle Protection Coveralls, Kimberly-Clark Professional
Supplier: Kimberly-Clark
KleenGuard®
49126 58534 49127 58515 58537 49006 49122 49005 58523 58506 49102 58505 58512 58525 49106 58526 58506 49112 49002 58514 49107 49115 58507 58517 49007 58536 58516 49103 49122 58502 58534 49105 58533 49004 58517 58527 49116 58524 49125 58503 58525 49003 49127 58535 49106 49005 58535 58504 49104 49114 58532 49126 58513 49002 49124 49124 49103 58526 49112 58514 58536 49107 58522 58504 58527 49117 58533 49113 49115 58523 58502 58515 58537 49105 58524 49004 58532 49123 58507 49114 49003 58503 58522 58505 49125
10815-966CS 381.34 USD InStock
10815-966 10815-968 CA82007-998 10815-964 10815-960 CA94001-702 10837-386 CA94001-704 CA10837-313 89030-740 CA94001-700 CA10837-316 CA10837-314 CA94001-706 CA10837-318 CA94001-708 CA83007-004 CA10815-704 CA10815-706 82004-880 CA83007-002 82004-882 94001-710 CA94001-714 CA10837-320 94001-704 94001-706 CA94001-710 10837-314 CA10837-324 CA94001-712 94001-702 CA10837-322 CA82004-882 10837-318 82004-872 CA10837-328 82004-874 10837-316 CA10837-326 82004-876 94001-708 82004-878 10815-706 83007-002 10815-702 CA82004-880 10815-704 83007-004 CA10815-992 CA94001-696 CA10837-382 CA10837-380 CA82004-878 10837-322 94001-712 CA10837-378 10837-326 94001-714 CA10837-332 10837-328 CA82004-872 CA82004-874 CA82004-876 CA10815-968 CA10815-964 CA10815-966 CA10815-960 94001-696 10837-380 82007-998 CA10815-962 94001-698 94001-692 10837-384 10837-382 94001-694 10837-332 CA10837-386 CA10837-384 CA94001-698 10837-378 CA10816-000 89030-738 89030-736
KleenGuard™ A20 Breathable Particle Protection Coveralls, Kimberly-Clark Professional
Disposable Garments
Made from a patented MICROFORCE™ barrier SMS fabric that offers barrier protection against dry particulates and light liquid sprays.
  • Seamless front provides more protection in the primary exposure areas
  • Zipper flap
The lightweight and breathable properties of the fabric make these coveralls comfortable to wear, while the strength and abrasion-resistant properties extend the use of this product. Patented Reflex™ Design is 7½ times less likely to rip out than ANSI minimums. Coveralls also provide 12% more chest room and 6% longer body length.
Certifications: NFPA 99 Compliant Antistatic Material.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty