Would you like to log in again?
Sklarlite XD™ Halsted Mosquito Forceps, OR Grade, Sklar
Supplier: Sklar
Sklarlite XD™
23-2505 23-2506
94002-744EA 158.64 USD InStock
94002-744 94002-742
Sklarlite XD™ Halsted Mosquito Forceps, OR Grade, Sklar
Forceps
Sklarlite XD™ Halsted Mosquito Forceps are are noticeably lighter and delicate than standard Halsted mosquito forceps, which allow the surgeon the highest degree of precision with the least amount of fatigue
The ratcheted finger ring handle provides a secure, locking grip to halt blood flow. They are not appropriate for use with large vessels or arteries. A toothed variation is available for grasping delicate tissue in skin grafting, biopsies, or ophthalmologic procedures. Mosquito forceps are known for their fine tips, and short, fully serrated jaws.
They are used in multiple procedures as a hemostat for small blood vessels.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty